siRNA Details
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108279.1 | NP_001101749.1 | 1 | 176 | ||
XM_008767841.1 | XP_008766063.1 | 2 | 271 | |||
XM_008767842.1 | XP_008766064.1 | 2 | 561 | |||
XM_008767843.1 | XP_008766065.1 | 2 | 261 | |||
GenBank mRNA | FQ223808.1 | - | 2 | 236 | ||
FQ224714.1 | - | 2 | 236 |
