siRNA Details
| Gene Symbol: RGD1308139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108281.1 | NP_001101751.1 | 12 | 1557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308139| Gene Symbol: RGD1308139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108281.1 | NP_001101751.1 | 12 | 1557 | ||