siRNA Details
| Gene Symbol: Spata22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191824.1 | NP_001178753.1 | 3, 4 | 242 | ||
| XM_006246762.2 | XP_006246824.1 | 4, 5 | 448 | |||
| XM_006246763.2 | XP_006246825.1 | 4, 5 | 342 | |||
| GenBank mRNA | AB236891.1 | - | 4, 5 | 436 | ||

Spata22
Genomic Map