siRNA Details
| Gene Symbol: Mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108289.1 | NP_001101759.1 | 3 | 303 | ||
| XM_008768075.1 | XP_008766297.1 | 3 | 494 | |||
| GenBank mRNA | FQ213501.1 | - | 3 | 839 | ||
| FQ216939.1 | - | 3 | 334 | |||

Mrps23
Genomic Map