siRNA Details
| Gene Symbol: Spata20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199402.1 | NP_955434.1 | 4 | 517 | ||
| XM_006247200.2 | XP_006247262.1 | 4 | 543 | |||
| GenBank mRNA | AY438568.1 | - | 4 | 517 | ||
| BC128718.1 | - | 4 | 477 | |||
| BC158571.1 | - | 4 | 476 | |||

Spata20
Genomic Map