siRNA Details
| Gene Symbol: Ormdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047897.1 | NP_001041362.1 | 3, 4 | 399 | ||
| XM_006247345.1 | XP_006247407.1 | 3, 4 | 835 | |||
| XM_006247346.1 | XP_006247408.1 | 3, 4 | 830 | |||
| GenBank mRNA | AY539934.1 | - | 3, 4 | 399 | ||

Ormdl3
Genomic Map