siRNA Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037202.1 | NP_001032279.1 | 6 | 1169 | ||
| XM_006247363.2 | XP_006247425.1 | 6 | 1156 | |||
| XM_006247364.2 | XP_006247426.1 | 6 | 1156 | |||
| XM_006247365.2 | XP_006247427.1 | 6 | 1230 | |||
| GenBank mRNA | BC107561.1 | - | 6 | 1169 | 7937018 | |

Plekhh3
Genomic Map