siRNA Details
| Gene Symbol: Dnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007726.1 | NP_001007727.1 | 7 | 1170 | ||
| GenBank mRNA | BC081920.1 | - | 7 | 1170 | 7112641 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnai2| Gene Symbol: Dnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007726.1 | NP_001007727.1 | 7 | 1170 | ||
| GenBank mRNA | BC081920.1 | - | 7 | 1170 | 7112641 | |