siRNA Details
Gene Symbol: Dnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007726.1 | NP_001007727.1 | 11, 12 | 1844 | ||
GenBank mRNA | BC081920.1 | - | 11, 12 | 1844 | 7112641 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007726.1 | NP_001007727.1 | 11, 12 | 1844 | ||
GenBank mRNA | BC081920.1 | - | 11, 12 | 1844 | 7112641 |