siRNA Details
| Gene Symbol: Cd300e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166577.1 | NP_001160049.1 | 4 | 611 | ||
| XM_008768394.1 | XP_008766616.1 | 5 | 937 | |||
| XM_008768395.1 | XP_008766617.1 | 5 | 941 | |||
| GenBank mRNA | GU057983.1 | - | 4 | 611 | ||

Cd300e
Genomic Map