siRNA Details
| Gene Symbol: Bahcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768494.1 | XP_008766716.1 | 20, 21 | 5757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bahcc1| Gene Symbol: Bahcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768494.1 | XP_008766716.1 | 20, 21 | 5757 | ||