siRNA Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108315.1 | NP_001101785.1 | 11, 12 | 1125 | ||
| XM_008768582.1 | XP_008766804.1 | 11, 12 | 1120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc3| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108315.1 | NP_001101785.1 | 11, 12 | 1125 | ||
| XM_008768582.1 | XP_008766804.1 | 11, 12 | 1120 | |||