siRNA Details
| Gene Symbol: RGD1310935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768658.1 | XP_008766880.1 | 3 | 1028 | ||
| XM_008768659.1 | XP_008766881.1 | 5 | 587 | |||
| XM_008768660.1 | XP_008766882.1 | 4 | 944 | |||
| XM_008768661.1 | XP_008766883.1 | 3 | 998 | |||
| GenBank mRNA | FQ229246.1 | - | 1 | 59 | ||

RGD1310935
Genomic Map