siRNA Details
| Gene Symbol: Popdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199113.1 | NP_954544.1 | 3 | 1048 | ||
| XM_008768733.1 | XP_008766955.1 | 4 | 3226 | |||
| XM_008768734.1 | XP_008766956.1 | 4 | 3226 | |||
| GenBank mRNA | AY490241.1 | - | 3 | 1011 | ||
| BC061876.1 | - | 3 | 1048 | 5599130 | ||

Popdc2
Genomic Map