siRNA Details
Gene Symbol: Rtp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108321.1 | NP_001101791.1 | 4 | 650 | ||
XM_006248521.2 | XP_006248583.1 | 2 | 1037 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rtp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108321.1 | NP_001101791.1 | 4 | 650 | ||
XM_006248521.2 | XP_006248583.1 | 2 | 1037 |