siRNA Details
Gene Symbol: Fam131a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727913.4 | XP_002727959.1 | 6 | 1065 | ||
XM_008768832.1 | XP_008767054.1 | 7 | 1329 | |||
XM_008768833.1 | XP_008767055.1 | 5 | 1027 | |||
XM_008768834.1 | XP_008767056.1 | 5 | 1124 |
Gene Symbol: LOC100912498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751082.3 | XP_003751130.1 | 1065 | |||
XM_008768817.1 | XP_008767039.1 | 1027 | ||||
XM_008768818.1 | XP_008767040.1 | 1124 | ||||
XM_008768819.1 | XP_008767041.1 | 788 |
