siRNA Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108329.1 | NP_001101799.1 | 2 | 302 | ||
| XM_006248835.2 | XP_006248897.1 | 2 | 429 | |||
| XM_008768976.1 | XP_008767198.1 | 1 | 5348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lnx2| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108329.1 | NP_001101799.1 | 2 | 302 | ||
| XM_006248835.2 | XP_006248897.1 | 2 | 429 | |||
| XM_008768976.1 | XP_008767198.1 | 1 | 5348 | |||