siRNA Details
Gene Symbol: Zfp655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008362.1 | NP_001008363.1 | 4 | 890 | ||
XM_006248895.2 | XP_006248957.1 | 4 | 984 | |||
XM_008768977.1 | XP_008767199.1 | 4 | 1256 | |||
XM_008768978.1 | XP_008767200.1 | 4 | 1345 | |||
GenBank mRNA | BC085937.1 | - | 4 | 890 | 7121505 |
Gene Symbol: LOC103690102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769057.1 | XP_008767279.1 | 3 | 799 |
