siRNA Details
| Gene Symbol: Zfp655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008362.1 | NP_001008363.1 | 4 | 1056 | ||
| XM_006248895.2 | XP_006248957.1 | 4 | 1150 | |||
| XM_008768977.1 | XP_008767199.1 | 4 | 1422 | |||
| XM_008768978.1 | XP_008767200.1 | 4 | 1511 | |||
| GenBank mRNA | BC085937.1 | - | 4 | 1056 | 7121505 | |
| Gene Symbol: LOC103690102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769057.1 | XP_008767279.1 | 3 | 965 | ||

Zfp655
Genomic Map