siRNA Details
Gene Symbol: Ccz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014126.1 | NP_001014148.1 | 7 | 656 | ||
XM_006248878.1 | XP_006248940.1 | 7 | 666 | |||
XM_006248879.1 | XP_006248941.1 | 7 | 666 | |||
GenBank mRNA | BC079086.1 | - | 7 | 656 | 7123188 |
