siRNA Details
| Gene Symbol: Ccz1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014126.1 | NP_001014148.1 | 3, 4 | 380 | ||
| XM_006248878.1 | XP_006248940.1 | 3, 4 | 390 | |||
| XM_006248879.1 | XP_006248941.1 | 3, 4 | 390 | |||
| GenBank mRNA | BC079086.1 | - | 3, 4 | 380 | 7123188 | |

Ccz1b
Genomic Map