siRNA Details
| Gene Symbol: Styxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037788.1 | NP_001032877.1 | 9 | 979 | ||
| XM_006249180.2 | XP_006249242.1 | 9 | 1202 | |||
| XM_006249181.2 | XP_006249243.1 | 9 | 1164 | |||
| XM_008769143.1 | XP_008767365.1 | 10 | 1357 | |||
| XM_008769144.1 | XP_008767366.1 | 8 | 1048 | |||
| XM_008769147.1 | XP_008767369.1 | 7 | 1016 | |||
| XM_008769148.1 | XP_008767370.1 | 7 | 974 | |||
| GenBank mRNA | BC098678.1 | - | 9 | 979 | 7382696 | |

Styxl1
Genomic Map