siRNA Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012147.1 | NP_001012147.1 | 9 | 1519 | ||
| XM_006249455.2 | XP_006249517.1 | 14 | 2941 | |||
| XM_006249457.2 | XP_006249519.1 | 11 | 2284 | |||
| XM_006249459.2 | XP_006249521.1 | 10 | 2181 | |||
| XM_006249460.2 | XP_006249522.1 | 10 | 1438 | |||
| GenBank mRNA | AY641810.1 | - | 10 | 1433 | ||
| BC081984.1 | - | 9 | 1519 | 7128801 | ||
| FQ226157.1 | - | 10 | 1434 | |||

Pxn
Genomic Map