siRNA Details
| Gene Symbol: Ankrd13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012148.1 | NP_001012148.1 | 6 | 838 | ||
| XM_006249461.1 | XP_006249523.1 | 6 | 1375 | |||
| XM_006249462.2 | XP_006249524.1 | 6 | 719 | |||
| GenBank mRNA | BC085774.1 | - | 6 | 838 | 7096477 | |

Ankrd13a
Genomic Map