siRNA Details
| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009448.1 | NP_001009448.1 | 2 | 404 | ||
| XM_006249653.2 | XP_006249715.1 | 3 | 1042 | |||
| GenBank mRNA | AB121032.1 | - | 2 | 404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh19| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009448.1 | NP_001009448.1 | 2 | 404 | ||
| XM_006249653.2 | XP_006249715.1 | 3 | 1042 | |||
| GenBank mRNA | AB121032.1 | - | 2 | 404 | ||