siRNA Details
| Gene Symbol: Scyl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191828.1 | NP_001178757.1 | 9 | 1307 | ||
| XM_006250175.2 | XP_006250237.1 | 7 | 764 | |||
| XM_006250176.2 | XP_006250238.1 | 8 | 789 | |||
| XM_008769660.1 | XP_008767882.1 | 8 | 2133 | |||
| XM_008769661.1 | XP_008767883.1 | 8 | 2133 | |||
| XM_008769662.1 | XP_008767884.1 | 9 | 1191 | |||

Scyl3
Genomic Map