siRNA Details
| Gene Symbol: Ccdc181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014131.1 | NP_001014153.1 | 2 | 266 | ||
| GenBank mRNA | BC078972.1 | - | 2 | 266 | 7111822 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc181| Gene Symbol: Ccdc181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014131.1 | NP_001014153.1 | 2 | 266 | ||
| GenBank mRNA | BC078972.1 | - | 2 | 266 | 7111822 | |