siRNA Details
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047103.1 | NP_001040568.1 | 3 | 320 | ||
| XM_006250323.2 | XP_006250385.1 | 4 | 592 | |||
| GenBank mRNA | BC161811.1 | - | 3 | 321 | 7317639 | |
| DQ272743.1 | - | 3 | 320 | |||

Cadm3
Genomic Map