siRNA Details
| Gene Symbol: Taf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037204.1 | NP_001032281.1 | 10, 11 | 1334 | ||
| XM_008769862.1 | XP_008768084.1 | 10, 11 | 1356 | |||
| GenBank mRNA | BC107466.1 | - | 10, 11 | 1334 | 7460374 | |

Taf1a
Genomic Map