siRNA Details
Gene Symbol: Rrp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009702.1 | NP_001009702.1 | 2, 3 | 412 | ||
GenBank mRNA | BC087649.1 | - | 2, 3 | 412 | 7310706 | |
FQ215627.1 | - | 2, 3 | 411 | |||
FQ219085.1 | - | 2, 3 | 433 | |||
FQ225526.1 | - | 2, 3 | 436 |
