siRNA Details
| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203336.2 | NP_976081.1 | 7 | 1401 | ||
| XM_006250443.2 | XP_006250505.1 | 6 | 2054 | |||
| GenBank mRNA | AB126962.1 | - | 6 | 1285 | ||
| AB126963.1 | - | 7 | 1513 | |||
| AY341057.1 | - | 6 | 1164 | |||

Esrrg
Genomic Map