siRNA Details
| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203336.2 | NP_976081.1 | 6 | 1114 | ||
| XM_006250443.2 | XP_006250505.1 | 5 | 1767 | |||
| GenBank mRNA | AB126962.1 | - | 5 | 998 | ||
| AB126963.1 | - | 6 | 1226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esrrg| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203336.2 | NP_976081.1 | 6 | 1114 | ||
| XM_006250443.2 | XP_006250505.1 | 5 | 1767 | |||
| GenBank mRNA | AB126962.1 | - | 5 | 998 | ||
| AB126963.1 | - | 6 | 1226 | |||