siRNA Details
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109376.2 | NP_001102846.1 | 3 | 472 | ||
| XM_008769875.1 | XP_008768097.1 | 4 | 530 | |||
| Gene Symbol: LOC317456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047894.1 | NP_001041359.1 | 2 | 73 | ||
| GenBank mRNA | AY383668.1 | - | 2 | 73 | ||

Lpgat1
Genomic Map