siRNA Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014132.1 | NP_001014154.1 | 7 | 722 | ||
| XM_008769865.1 | XP_008768087.1 | 6 | 1387 | |||
| XM_008769866.1 | XP_008768088.1 | 6 | 1387 | |||
| GenBank mRNA | BC089843.1 | - | 7 | 722 | 7368358 | |

Traf3ip3
Genomic Map