siRNA Details
| Gene Symbol: Fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199114.1 | NP_954545.1 | 2, 3 | 487 | ||
| GenBank mRNA | AJ536020.1 | - | 2, 3 | 487 | ||
| BC097261.1 | - | 2, 3 | 443 | 7442454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgfrl1| Gene Symbol: Fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199114.1 | NP_954545.1 | 2, 3 | 487 | ||
| GenBank mRNA | AJ536020.1 | - | 2, 3 | 487 | ||
| BC097261.1 | - | 2, 3 | 443 | 7442454 | ||