siRNA Details
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170456.1 | NP_001163927.1 | 5 | 870 | ||
GenBank mRNA | BC166752.1 | - | 5 | 821 | 5621368 | |
FQ211286.1 | - | 1059 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170456.1 | NP_001163927.1 | 5 | 870 | ||
GenBank mRNA | BC166752.1 | - | 5 | 821 | 5621368 | |
FQ211286.1 | - | 1059 |