siRNA Details
| Gene Symbol: Fam69a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170456.1 | NP_001163927.1 | 5 | 648 | ||
| GenBank mRNA | BC166752.1 | - | 5 | 599 | 5621368 | |
| FQ211286.1 | - | 837 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam69a| Gene Symbol: Fam69a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170456.1 | NP_001163927.1 | 5 | 648 | ||
| GenBank mRNA | BC166752.1 | - | 5 | 599 | 5621368 | |
| FQ211286.1 | - | 837 | ||||