siRNA Details
Gene Symbol: Tmem150c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108354.2 | NP_001101824.1 | 3, 4 | 179 | ||
XM_006250670.2 | XP_006250732.1 | 3, 4 | 514 | |||
XM_006250671.2 | XP_006250733.1 | 3, 4 | 2203 | |||
GenBank mRNA | BC169066.1 | - | 3, 4 | 179 | 5598025 | |
FQ214560.1 | - | 3, 4 | 196 |
