siRNA Details
| Gene Symbol: Cnot6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108355.1 | NP_001101825.1 | 10, 11 | 1388 | ||
| XM_006250708.2 | XP_006250770.1 | 10, 11 | 1420 | |||
| XM_006250709.2 | XP_006250771.1 | 10, 11 | 1307 | |||
| XM_008770042.1 | XP_008768264.1 | 9, 10 | 4187 | |||
| XM_008770043.1 | XP_008768265.1 | 9, 10 | 1199 | |||

Cnot6l
Genomic Map