siRNA Details
| Gene Symbol: Zcchc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108360.1 | NP_001101830.1 | 6 | 822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc4| Gene Symbol: Zcchc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108360.1 | NP_001101830.1 | 6 | 822 | ||