siRNA Details
| Gene Symbol: Usp34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271196.1 | NP_001258125.1 | 77 | 9807 | ||
| GenBank mRNA | BC107808.1 | - | 4 | 246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp34| Gene Symbol: Usp34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271196.1 | NP_001258125.1 | 77 | 9807 | ||
| GenBank mRNA | BC107808.1 | - | 4 | 246 | ||