siRNA Details
| Gene Symbol: Arhgef40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271313.1 | NP_001258242.1 | 4 | 1476 | ||
| XM_008770553.1 | XP_008768775.1 | 4 | 1538 | |||
| XM_008770554.1 | XP_008768776.1 | 4 | 1538 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef40| Gene Symbol: Arhgef40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271313.1 | NP_001258242.1 | 4 | 1476 | ||
| XM_008770553.1 | XP_008768775.1 | 4 | 1538 | |||
| XM_008770554.1 | XP_008768776.1 | 4 | 1538 | |||