siRNA Details
| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012155.1 | NP_001012155.1 | 1 | 357 | ||
| XM_008770688.1 | XP_008768910.1 | 1 | 414 | |||
| GenBank mRNA | BC081891.1 | - | 1 | 357 | 7109579 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm9sf1| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012155.1 | NP_001012155.1 | 1 | 357 | ||
| XM_008770688.1 | XP_008768910.1 | 1 | 414 | |||
| GenBank mRNA | BC081891.1 | - | 1 | 357 | 7109579 | |