siRNA Details
| Gene Symbol: Ccdc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108382.1 | NP_001101852.1 | 4, 5 | 283 | ||
| XM_006252132.2 | XP_006252194.1 | 4, 5 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc25| Gene Symbol: Ccdc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108382.1 | NP_001101852.1 | 4, 5 | 283 | ||
| XM_006252132.2 | XP_006252194.1 | 4, 5 | 282 | |||