siRNA Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108383.1 | NP_001101853.1 | 8 | 1547 | ||
| XM_006252133.2 | XP_006252195.1 | 7 | 1124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ebf2| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108383.1 | NP_001101853.1 | 8 | 1547 | ||
| XM_006252133.2 | XP_006252195.1 | 7 | 1124 | |||