siRNA Details
| Gene Symbol: Sucla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108387.2 | NP_001101857.2 | 6 | 757 | ||
| GenBank mRNA | BC166998.1 | - | 5 | 677 | ||
| CF110427.1 | - | 2 | 183 | |||
| FQ216292.1 | - | 2 | 123 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sucla2| Gene Symbol: Sucla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108387.2 | NP_001101857.2 | 6 | 757 | ||
| GenBank mRNA | BC166998.1 | - | 5 | 677 | ||
| CF110427.1 | - | 2 | 183 | |||
| FQ216292.1 | - | 2 | 123 | |||