siRNA Details
| Gene Symbol: Cog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012157.1 | NP_001012157.1 | 4 | 607 | ||
| XM_006252323.2 | XP_006252385.1 | 4 | 1200 | |||
| GenBank mRNA | BC083901.1 | - | 4 | 607 | 7133573 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cog3| Gene Symbol: Cog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012157.1 | NP_001012157.1 | 4 | 607 | ||
| XM_006252323.2 | XP_006252385.1 | 4 | 1200 | |||
| GenBank mRNA | BC083901.1 | - | 4 | 607 | 7133573 | |