siRNA Details
| Gene Symbol: Cog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012157.1 | NP_001012157.1 | 8, 9 | 1001 | ||
| XM_006252323.2 | XP_006252385.1 | 8, 9 | 1594 | |||
| GenBank mRNA | BC083901.1 | - | 8, 9 | 1001 | 7133573 | |

Cog3
Genomic Map