siRNA Details
Gene Symbol: Rnf219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108389.1 | NP_001101859.1 | 1 | 47 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026047, AK013314, BI986775.
Gene Ontology Categories:
