siRNA Details
| Gene Symbol: Top2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100858.1 | NP_001094328.1 | 24 | 3974 | ||
| GenBank mRNA | AB262979.1 | - | 24 | 3974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Top2b| Gene Symbol: Top2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100858.1 | NP_001094328.1 | 24 | 3974 | ||
| GenBank mRNA | AB262979.1 | - | 24 | 3974 | ||